2-([1,1':4',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%

Supplier: Ambeed
Warning

Synonyms: (1,1':4',1''-Terphenyl)-4-ylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(1,1':4',1''-terphenyl)-4-yl-1,3,2-dioxaborolane

A254301-1G A254301-250MG A254301-25G A254301-5G
76976-742EA 56.23 USD
76976-742 76976-744 76976-746 76976-748
2-([1,1':4',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-([1,1':4',1''-Terphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₄H₂₅BO₂
MW: 356.27 g/mol
MDL Number: MFCD16294429
CAS Number: 1080632-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR