Thieno[2,3-d]pyrimidine, 4-chloro-6-(1,1-dimethylethyl)- 95%

Supplier: Ambeed
Warning

A330203-100MG A330203-1G A330203-250MG
76978-224EA 41.38 USD
76978-224 76978-226 76978-228
Thieno[2,3-d]pyrimidine, 4-chloro-6-(1,1-dimethylethyl)- 95%
Thieno[2,3-d]pyrimidine, 4-chloro-6-(1,1-dimethylethyl)-

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂S
MW: 226.72 g/mol
MDL Number: MFCD04973932
CAS Number: 439692-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR