Methyl 2-amino-5-tert-butylthiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

A696840-100MG A696840-1G A696840-250MG
76981-056EA 10 USD
76981-056 76981-058 76981-060
Methyl 2-amino-5-tert-butylthiophene-3-carboxylate 97%
Methyl 2-amino-5-tert-butylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MDL Number: MFCD08059073
CAS Number: 216574-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR