1-Isopropyl-4-piperidone 97%

Supplier: Ambeed
Warning

A147972-100G A147972-10G A147972-1G A147972-25G A147972-5G
76972-922EA 106 USD
76972-922 76972-924 76972-926 76972-928 76972-930
1-Isopropyl-4-piperidone 97%
1-Isopropyl-4-piperidone

Precaución: for Research Use Only

Formula: C₈H₁₅NO
MDL Number: MFCD00038035
CAS Number: 5355-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR