3-Bromo-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A128142-100MG A128142-250MG A128142-10G A128142-1G A128142-25G
76971-954EA 6 USD
76971-954 76971-956 77611-116 77670-818 77881-027
3-Bromo-2-(trifluoromethyl)pyridine 98%
3-Bromo-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C6H3BrF3N
MW: 225.9939 g/mol
MDL Number: MFCD08458124
CAS Number: 590371-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR