(1H-Pyrrol-2-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A384505-100MG A384505-1G A384505-250MG A384505-5G
76978-858EA 25 USD
76978-858 76978-860 76978-862 77672-908
(1H-Pyrrol-2-yl)methanamine hydrochloride 95%
(1H-Pyrrol-2-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂
MDL Number: MFCD20037390
CAS Number: 1351479-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR