Cyclopentylmethanamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Aminomethylcyclopentane hydrochloride

A115606-1G A115606-250MG A115606-5G A115606-25G
76971-134EA 29 USD
76971-134 76971-136 76971-138 77566-078
Cyclopentylmethanamine hydrochloride 97%
Cyclopentylmethanamine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₄ClN
MDL Number: MFCD07403659
CAS Number: 58714-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR