Ethyl 4,6-dichloropyridazine-3-carboxylate 95%

Supplier: Ambeed
Warning

A267260-1G A267260-5G A267260-25G A267260-250MG
76977-132EA 87 USD
76977-132 76977-134 77571-656 77612-566
Ethyl 4,6-dichloropyridazine-3-carboxylate 95%
Ethyl 4,6-dichloropyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.04 g/mol
MDL Number: MFCD11521570
CAS Number: 679406-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR