(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Fluoropyrrolidine hydrochloride

A214432-100G A214432-10G A214432-1G A214432-250MG A214432-25G A214432-5G
76975-620EA 1156.39 USD
76975-620 76975-622 76975-624 76975-626 76975-628 76975-630
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClFN
MW: 125.57 g/mol
MDL Number: MFCD04038718
CAS Number: 136725-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR