Methyl-6-oxopiperidine-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-oxopiperidine-3-carboxylate
A375446-100MG
A375446-250MG
A375446-1G
76978-766EA
26.53
USD
76978-766
76978-768
77594-128
Methyl-6-oxopiperidine-3-carboxylate 95%
Methyl-6-oxopiperidine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₁NO₃ MW: 157.17 g/mol |
MDL Number:
MFCD17016096 CAS Number: 958991-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...