Tetrachlorophthalonitrile 97%

Supplier: Ambeed
Warning

A291600-100G A291600-25G A291600-500G A291600-5G
76977-580EA 54.11 USD
76977-580 76977-582 76977-584 76977-586
Tetrachlorophthalonitrile 97%
Tetrachlorophthalonitrile

Precaución: for Research Use Only

Formula: C₈Cl₄N₂
MDL Number: MFCD00019740
CAS Number: 1953-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR