tert-Butyl-4-(isopropylamino)piperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(isopropylamino)piperidine

A915900-100MG A915900-250MG A915900-5G A915900-1G
76981-950EA 37 USD
76981-950 76981-952 76981-954 77308-516
tert-Butyl-4-(isopropylamino)piperidine-1-carboxylate 96%
tert-Butyl-4-(isopropylamino)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₆N₂O₂
MDL Number: MFCD07367732
CAS Number: 534595-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR