(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-methyl-1H-indol-3-yl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Trp(2-Me)-OH

A367212-100MG A367212-1G A367212-250MG A367212-5G
76978-648EA 52 USD
76978-648 76978-650 76978-652 77881-996
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-methyl-1H-indol-3-yl)propanoic acid 95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-methyl-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MW: 440.49 g/mol
MDL Number: MFCD07366896
CAS Number: 196808-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR