(S)-3-Methylmorpholine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A213493-1G
A213493-250MG
A213493-25G
76975-596EA
11
USD
76975-596
76975-598
76975-600
(S)-3-Methylmorpholine hydrochloride 97%
(S)-3-Methylmorpholine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁NO·ClH MW: 137.61 g/mol |
MDL Number:
MFCD18382512 CAS Number: 1022094-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...