(E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one 97%
Supplier: Ambeed
|
Warning
|
A123800-100G
A123800-10G
A123800-1G
A123800-25G
A123800-5G
76971-702EA
95.49
USD
76971-702
76971-704
76971-706
76971-708
76971-710
(E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one 97%
(E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₂O MW: 176.22 g/mol |
MDL Number:
MFCD00115180 CAS Number: 123367-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...