Methyl 3-oxo-3-(pyridin-2-yl)propanoate 95%

Supplier: Ambeed
Warning

A150317-10G A150317-1G A150317-250MG A150317-5G
76973-066EA 395.72 USD
76973-066 76973-068 76973-070 76973-072
Methyl 3-oxo-3-(pyridin-2-yl)propanoate 95%
Methyl 3-oxo-3-(pyridin-2-yl)propanoate

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.173 g/mol
MDL Number: MFCD11849520
CAS Number: 75418-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR