[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid 98+%
Supplier: Ambeed
|
Warning
|
A744905-100MG
A744905-1G
A744905-250MG
76981-336EA
46.68
USD
76981-336
76981-338
76981-340
[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid 98+%
[1,2,4]triazolo[1,5-a]pyridine-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅N₃O₂ MW: 163.13 g/mol |
MDL Number:
MFCD12923316 CAS Number: 1043903-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...