Palladium(II) iodide 98%

Supplier: Ambeed
Warning

A251126-1G A251126-250MG A251126-5G
76976-688EA 75 USD
76976-688 77881-671 77881-672
Palladium(II) iodide 98%
Palladium(II) iodide

Precaución: for Research Use Only

Formula: I₂Pd
MW: 360.23
Density: 6.003
MDL Number: MFCD00011171
CAS Number: 7790-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR