(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A272222-100G A272222-10G A272222-1G A272222-250MG A272222-25G A272222-5G
76977-196EA 1368.57 USD
76977-196 76977-198 76977-200 76977-202 76977-204 76977-206
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD03094727
CAS Number: 72925-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR