6-Aminobenzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-1,3-benzothiazole, Benzothiazol-6-ylamine

A161652-10G A161652-250MG A161652-5G A161652-1G A161652-25G A161652-100G
76973-614EA 102.91 USD
76973-614 76973-616 76973-618 77567-358 77611-570 77671-290
6-Aminobenzothiazole 98%
6-Aminobenzothiazole

Precaución: For research use only.

Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD00015461
CAS Number: 533-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR