N-(Bromomethyl)phthalimide 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Bromomethyl)isoindoline-1,3-dione

A175810-100G A175810-10G A175810-1G A175810-25G A175810-500G A175810-5G
76974-242EA 76 USD
76974-242 76974-244 76974-246 76974-248 76974-250 76974-252
N-(Bromomethyl)phthalimide 97%
N-(Bromomethyl)phthalimide

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD00005897
CAS Number: 5332-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR