(3-Amino-1H-pyrazol-4-yl)(thiophen-2-yl)methanone 95%

Supplier: Ambeed
Warning

A222869-100MG A222869-1G A222869-250MG A222869-5G
76975-888EA 20.16 USD
76975-888 76975-890 76975-892 76975-894
(3-Amino-1H-pyrazol-4-yl)(thiophen-2-yl)methanone 95%
(3-Amino-1H-pyrazol-4-yl)(thiophen-2-yl)methanone

Precaución: for Research Use Only

Formula: C₈H₇N₃OS
MW: 193.22 g/mol
MDL Number: MFCD20260223
CAS Number: 96219-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR