3-Bromo-4-iodopyridine 97%

Supplier: Ambeed
Warning

A165947-1G A165947-250MG A165947-5G A165947-10G
76973-784EA 18 USD
76973-784 76973-786 76973-788 77671-330
3-Bromo-4-iodopyridine 97%
3-Bromo-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
MDL Number: MFCD11110263
CAS Number: 89167-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR