1-Methyl-7-azaindole-4-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A597871-100MG
A597871-1G
A597871-250MG
76980-486EA
31.83
USD
76980-486
76980-488
76980-490
1-Methyl-7-azaindole-4-carboxylic acid 95%
1-Methyl-7-azaindole-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ |
MDL Number:
MFCD13302392 CAS Number: 1147753-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...