1H-Benzimidazole-2-sulfonic acid 95%

Supplier: Ambeed
Danger

Synonyms: 1H-Benzo[d]imidazole-2-sulphonic acid

A574997-1G A574997-250MG A574997-25G A574997-5G
76980-352EA 27.59 USD
76980-352 76980-354 76980-356 76980-358
1H-Benzimidazole-2-sulfonic acid 95%
1H-Benzimidazole-2-sulphonic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₃S
MW: 198.1991 g/mol
MDL Number: MFCD00100972
CAS Number: 40828-54-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR