(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-2-Amino-1-(Boc-amino)cyclohexane, (1R,2S)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, tert-Butyl (1R,2S)-2-aminocyclohexylcarbamate, (1R,2S)-N1-Boc-1,2-cyclohexanediamine

A683979-250MG A683979-5G A683979-1G
76981-016EA 35.01 USD
76981-016 77614-116 77673-868
(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 98%
(1R,2S)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD09952106
CAS Number: 364385-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR