(1-(4-Nitrophenyl)piperidin-3-yl)methanol 97%

Supplier: Ambeed
Warning

A412324-1G A412324-5G
76979-166EA 46 USD
76979-166 76979-168
(1-(4-Nitrophenyl)piperidin-3-yl)methanol 97%
(1-(4-Nitrophenyl)piperidin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD11039478
CAS Number: 166438-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR