7-Bromo-4-methylquinoline 98%

Supplier: Ambeed
Warning

A275216-100MG A275216-1G A275216-250MG A275216-5G
76977-280EA 8 USD
76977-280 76977-282 76977-284 76977-286
7-Bromo-4-methylquinoline 98%
7-Bromo-4-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD18204665
CAS Number: 141052-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR