Boc-D-asp-Otbu 97%

Supplier: Ambeed
Warning

A237707-1G A237707-250MG A237707-10G A237707-25G A237707-100G A237707-5G
76976-374EA 11 USD
76976-374 76976-376 77593-522 77593-524 77881-621 77881-622
Boc-D-asp-Otbu 97%
Boc-D-asp-Otbu

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₆
MDL Number: MFCD00236827
CAS Number: 77004-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR