RU-58841 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
RU 58841, RU58841, 4-[3-(4-hydroxybutyl)-4,4-dimethyl-2,5-dioxoimidazolidin-1-yl]-2-(trifluoromethyl)benzonitrile, 4-(3-(4-Hydroxybutyl)-4,4-dimethyl-2,5-dioxoimidazolidin-1-yl)-2-(trifluoromethyl)benzonitrile
A108339-100MG
A108339-10MG
A108339-50MG
A108339-5MG
A108339-5G
A108339-1G
A108339-25G
76970-554EA
20
USD
76970-554
76970-556
76970-558
76970-560
77570-482
77880-874
77880-875
RU-58841 98%
RU-58841
Precaución: for Research Use Only
Formula:
C₁₇H₁₈F₃N₃O₃ MW: 369.34 g/mol |
MDL Number:
MFCD00925776 CAS Number: 154992-24-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...