2-Chloro-6-naphthol 98%

Supplier: Ambeed
Warning

A323363-1G A323363-250MG
76978-094EA 111 USD
76978-094 76978-096
2-Chloro-6-naphthol 98%
2-Chloro-6-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇ClO
MDL Number: MFCD00127706
CAS Number: 40604-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR