4-Chloro-6-methyl-2-trifluoromethylpyrimidine 98%

Supplier: Ambeed
Warning

A653489-100MG A653489-1G A653489-250MG
76980-832EA 48.81 USD
76980-832 77594-988 77673-778
4-Chloro-6-methyl-2-trifluoromethylpyrimidine 98%
4-Chloro-6-methyl-2-trifluoromethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClF₃N₂
MW: 196.558 g/mol
MDL Number: MFCD09743686
CAS Number: 1582-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR