3-Chloro-4,5-dimethoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A645360-250MG A645360-1G A645360-5G
76980-774EA 22 USD
76980-774 77673-752 77673-754
3-Chloro-4,5-dimethoxybenzaldehyde 95%
3-Chloro-4,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD02256335
CAS Number: 18268-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR