1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4-Dihydronaphthalen-1(2H)-one
A290189-1000G
A290189-100G
A290189-10G
A290189-25G
A290189-500G
76977-548EA
138.98
USD
76977-548
76977-550
76977-552
76977-554
76977-556
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) 97%
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone)
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O MW: 146.19 g/mol |
MDL Number:
MFCD00001688 CAS Number: 529-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...