5-Chloro-2,3-diphenyl-1H-indole 97%

Supplier: Ambeed
Warning

A241957-1G A241957-250MG A241957-5G
76976-488EA 23 USD
76976-488 76976-490 76976-492
5-Chloro-2,3-diphenyl-1H-indole 97%
5-Chloro-2,3-diphenyl-1H-indole

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClN
MDL Number: MFCD00974243
CAS Number: 52598-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR