9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-Phenylcarbazole-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-3-yl)-1,3,2-dioxaborolane, 9-Phenyl-9H-carbazol-3-yl-3-boronic acid pinacol ester, 9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
A121340-100G
A121340-10G
A121340-1G
A121340-25G
A121340-500G
A121340-5G
76971-530EA
389.36
USD
76971-530
76971-532
76971-534
76971-536
76971-538
76971-540
9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 97%
9-Phenyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Precaución: for Research Use Only
Formula:
C₂₄H₂₄BNO₂ MW: 369.27 g/mol |
MDL Number:
MFCD16621140 CAS Number: 1126522-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...