3-Hydroxy-4-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A130163-100G A130163-10G A130163-25G A130163-5G
76972-022EA 59 USD
76972-022 76972-024 76972-026 76972-028 76972-030
3-Hydroxy-4-methoxybenzonitrile 97%
3-Hydroxy-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.1500 g/mol
MDL Number: MFCD02258854
CAS Number: 52805-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR