trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: trans-Methyl 4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

A165612-100MG A165612-1G A165612-250MG A165612-5G A165612-25G A165612-100G
76973-772EA 5 USD
76973-772 76973-774 76973-776 76973-778 77567-408 77881-283
trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate 97%
trans-Methyl-4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

Precaución: For research use only.

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD17015041
CAS Number: 146307-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR