6-Chlorobenzo[d]thiazole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A916309-100MG A916309-1G A916309-250MG
76981-956EA 77 USD
76981-956 76981-958 76981-960
6-Chlorobenzo[d]thiazole-2-carbonitrile 95%
6-Chlorobenzo[d]thiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClN₂S
MDL Number: MFCD11217322
CAS Number: 26649-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR