2-Hydroxy-4-methoxybenzonitrile 97%

Supplier: Ambeed
Warning

A587584-10G A587584-1G A587584-250MG A587584-5G
76980-428EA 279.02 USD
76980-428 76980-430 76980-432 76980-434
2-Hydroxy-4-methoxybenzonitrile 97%
2-Hydroxy-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.1500 g/mol
MDL Number: MFCD08567959
CAS Number: 39835-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR