1-Tritylimidazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Trityl-1H-imidazole-4-carboxylic acid

A697283-1G A697283-250MG
76981-062EA 111.4 USD
76981-062 76981-064
1-Tritylimidazole-4-carboxylic acid 98%
1-Tritylimidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₃H₁₈N₂O₂
MW: 354.4 g/mol
MDL Number: MFCD06203351
CAS Number: 191103-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR