3-Bromo-2-chloro-6-methyl-5-nitropyridine 98%

Supplier: Ambeed
Danger

A196435-100MG A196435-10G A196435-250MG A196435-25G A196435-5G
76975-070EA 5 USD
76975-070 76975-072 76975-074 76975-076 76975-078
3-Bromo-2-chloro-6-methyl-5-nitropyridine 98%
3-Bromo-2-chloro-6-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MW: 251.47 g/mol
MDL Number: MFCD09839270
CAS Number: 856834-95-2
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR