2,4-Dichloro-3,6-dimethylpyridine 95%

Supplier: Ambeed
Warning

A304765-1G A304765-5G A304765-25G
76977-778EA 28 USD
76977-778 77672-544 77881-843
2,4-Dichloro-3,6-dimethylpyridine 95%
2,4-Dichloro-3,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MDL Number: MFCD11113423
CAS Number: 83791-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR