Ethyl [1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylate 95%

Supplier: Ambeed
Warning

A107740-100MG A107740-250MG
76970-536EA 51.99 USD
76970-536 76970-538
Ethyl [1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylate 95%
Ethyl [1,2,4]triazolo[4,3-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈N₄O₂
MW: 192.18 g/mol
MDL Number: MFCD18338245
CAS Number: 1335055-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR