7-Bromoquinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

A195941-100MG A195941-1G A195941-5G
76975-034EA 13 USD
76975-034 76975-036 76975-038
7-Bromoquinolin-2(1H)-one 98%
7-Bromoquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD09743441
CAS Number: 99465-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR