4-Bromoisoquinolin-3-amine 97%

Supplier: Ambeed
Warning

A478424-100MG A478424-1G A478424-250MG
76979-720EA 11 USD
76979-720 76979-722 76979-724
4-Bromoisoquinolin-3-amine 97%
4-Bromoisoquinolin-3-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD00176837
CAS Number: 10321-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR