Phenoxybenzamine hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Dibenzyline Hydrochloride, N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine Hydrochloride, N-Phenoxyisopropyl-N-benzyl-2-chloroethylamine Hydrochloride, N-Benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine hydrochloride

A398762-1G A398762-1MG A398762-250MG A398762-25G A398762-5G
76979-024EA 34 USD
76979-024 76979-026 76979-028 76979-030 76979-032
Phenoxybenzamine hydrochloride 98%
Phenoxybenzamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₂ClNO
MW: 340.29 g/mol
MDL Number: MFCD00055152
CAS Number: 63-92-3
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR