Methyl-2-amino-3-hydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-3-hydroxybenzoate

A608394-100MG A608394-1G A608394-250MG A608394-5G
76980-548EA 24.41 USD
76980-548 76980-550 76980-552 76980-554
Methyl-2-amino-3-hydroxybenzoate 98%
Methyl-2-amino-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD01675233
CAS Number: 17672-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR