Ethyl-4-chloro-2-methoxypyrimidine-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-chloro-2-methoxypyrimidine-5-carboxylate
A208437-1G
A208437-5G
76975-426EA
81.69
USD
76975-426
76975-428
Ethyl-4-chloro-2-methoxypyrimidine-5-carboxylate 98%
Ethyl-4-chloro-2-methoxypyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₉ClN₂O₃ MW: 216.6217 g/mol |
MDL Number:
MFCD12923009 CAS Number: 65269-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...