3-Carbamoylbenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A111256-1G
A111256-100MG
A111256-250MG
A111256-5G
76970-796EA
50
USD
76970-796
77670-574
77670-576
77880-897
3-Carbamoylbenzoic acid 95%
3-Carbamoylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇NO₃ |
MDL Number:
MFCD04118098 CAS Number: 4481-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...